Learn More
Kingfisher Biotech Inc Zebrafish VEGF-A ELISpot - Up to two (2) ELISpot assays
SDP
Catalog No. 502891871
Encompass_Preferred
Zebrafish VEGF-A ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA.
Catalog No. 50-289-1871 Supplier Kingfisher Biotech Inc Supplier No. SPOT2570Z004
Zebrafish VEGF-A ELISpot contains Capture Antibody, Standard Control, and Detection Antibody for development of up to two (2) ELISpot assays. Other required materials and reagents are not supplied. The antibodies have been determined to function in an ELISpot with the Standard Control provided. Components may also be purchased separately. For research use only. Made in the USA.