Learn More
Phenomenex™ Zebron Gas Management™ Connecting Units for Gas Management Filters, 1-Position, GC-MSD (Universal) Detector
Shop All Phenomenex Products
Description
- Product Type: Connecting Unit
- Format: 1-Position
- For Use With: Gas Management Filters
- Dectector Type: GC-MSD (Universal)
Specifications
Specifications
| For Use With (Equipment) | Click-On Inline Filter |
| Diameter (Metric) Inner | 3.2 mm |
| Material | Stainless Steel |
| Technique | Gas Chromatography |
| Type | Double-ended Connector |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.