Description
- Attach a new filter in under 10 seconds
- Optional electronic indicator display
- Stainless steel to withstand high pressures
- Secure wall-mounting using clamps
Specifications
Specifications
| Cartridge Type | Moisture |
| For Use With (Equipment) | Click-On Inline Filter |
| Indicator Change | Yellow to Clear |
| Technique | Gas Chromatography |
| Type | Indicating Moisture Cartridge Filter |