Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.53 mm x 1.0 m
Catalog No. 502335357 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-624 7HK-G005-22
Catalog No. 50-233-5357 Supplier Phenomenex Inc Supplier No. 7HKG00522
Zebron™ ZB-624 7HK-G005-22