Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.25 mm x 0.10 m
Catalog No. 502335661 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-1HT 7HG-G014-02
Catalog No. 50-233-5661 Supplier Phenomenex Inc Supplier No. 7HGG01402
Zebron™ ZB-1HT 7HG-G014-02