Learn More
Phenomenex Inc GC Cap. Column 60 m x 0.25 mm x 0.25 m
Catalog No. 502335670 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-1plus 7KG-G031-11
Catalog No. 50-233-5670 Supplier Phenomenex Inc Supplier No. 7KGG03111
Zebron™ ZB-1plus 7KG-G031-11