Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.53 mm x 2.00 m
Catalog No. 502335689 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-BAC-2 7HK-G022-32
Catalog No. 50-233-5689 Supplier Phenomenex Inc Supplier No. 7HKG02232
Zebron™ ZB-BAC-2 7HK-G022-32