Learn More
Phenomenex Inc GC Cap. Column 60 m x 0.53 mm x 3.00 m
Catalog No. 502335702 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-624 7KK-G005-36
Catalog No. 50-233-5702 Supplier Phenomenex Inc Supplier No. 7KKG00536
Zebron™ ZB-624 7KK-G005-36