Learn More
Phenomenex Inc GC Cap. Column 60m x 0.25mm x 1.4um
Catalog No. 502335734 Shop All Phenomenex Products
Encompass_Preferred
Zebron ZB-624plus 7KG-G040-27
Catalog No. 50-233-5734 Supplier Phenomenex Inc Supplier No. 7KGG04027
Zebron ZB-624plus 7KG-G040-27