Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc GC Cap. Column 15 m x 0.32 mm x 0.10 m

Catalog No. 502335402 Shop All Phenomenex Products
Encompass_Preferred

Zebron™ ZB-5HT w/Spliced Guard 2 m 7EM-G015-02-GST

Catalog No. 50-233-5402 Supplier Phenomenex Inc Supplier No. 7EMG01502GST
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zebron™ ZB-5HT w/Spliced Guard 2 m 7EM-G015-02-GST

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.