Learn More
Phenomenex Inc GC Cap. Column 5m x 0.25mm x 1.00um
Catalog No. 502335475 Shop All Phenomenex Products
Encompass_Preferred
Zebron ZB-5 7AG-G002-22
Catalog No. 50-233-5475 Supplier Phenomenex Inc Supplier No. 7AGG00222
Zebron ZB-5 7AG-G002-22