Learn More
Phenomenex Inc GC Cap. Column 60m x 0.53mm x 3.0um
Catalog No. 502335501 Shop All Phenomenex Products
Encompass_Preferred
Zebron ZB-624plus 7KK-G040-36
Catalog No. 50-233-5501 Supplier Phenomenex Inc Supplier No. 7KKG04036
Zebron ZB-624plus 7KK-G040-36