Learn More
Phenomenex Inc GC Cap. Column 100 m x 0.25 mm x 0.50 m
Catalog No. 502335518 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-1 7MG-G001-17
Catalog No. 50-233-5518 Supplier Phenomenex Inc Supplier No. 7MGG00117
Zebron™ ZB-1 7MG-G001-17