Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc GC Column Dual Integrated Guard, 1.0m on Each End

Catalog No. 502335532 Shop All Phenomenex Products
Encompass_Preferred

Zebron™ ZB-1HT 2m x 0.25mm x 0.10m, With CG0-9347

Catalog No. 50-233-5532 Supplier Phenomenex Inc Supplier No. CG09347
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zebron™ ZB-1HT 2m x 0.25mm x 0.10m, With CG0-9347

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.