Learn More
Phenomenex Inc Zebron ZB-5MS
Catalog No. 502335547 Shop All Phenomenex Products
Encompass_Preferred
Zebron ZB-5MS GC Cap. Column 20 m x 0.18 mm x 0.32 µm
Catalog No. 50-233-5547 Supplier Phenomenex Inc Supplier No. 7FDG01051
Zebron ZB-5MS GC Cap. Column 20 m x 0.18 mm x 0.32 µm