Learn More
Phenomenex Inc Zebron ZB-5 w/GUARDIAN, 5 m, GC Capillary Column 15 m x 0.25 mm x 0.25 µm, Ea
Supplier:  Phenomenex Inc 7EGG00211GGA
Encompass
95% Dimethylpolysiloxane 5% Pheny low polarity phase for general purpose useRecommended Applications: Alkaloids, Dioxins, Drugs, Essential Oils/Flavors, Halo-hydrocarbons, PCBs/Aroclors, Pesticides/Herbicides, Phenols, Residual Solvents, Semi-volatiles
Catalog No. 50-233-5561
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.