Learn More
Phenomenex Inc GC Cap. Column 10m x 0.25mm x 4um
Catalog No. 502335568 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-1 7CG-G001-37
Catalog No. 50-233-5568 Supplier Phenomenex Inc Supplier No. 7CGG00137
Zebron™ ZB-1 7CG-G001-37