Learn More
Phenomenex Inc GC Cap. Column 60 m x 0.32 mm x 1.0 m
Catalog No. 502335690 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-5MSplus 7KM-G030-22
Catalog No. 50-233-5690 Supplier Phenomenex Inc Supplier No. 7KMG03022
Zebron™ ZB-5MSplus 7KM-G030-22