Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.25 mm x 0.25 m
Catalog No. 502335391 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-5HT 7HG-G015-11
Catalog No. 50-233-5391 Supplier Phenomenex Inc Supplier No. 7HGG01511
Zebron™ ZB-5HT 7HG-G015-11