Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.53 mm x 0.50 m
Catalog No. 502335433 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-5 7HK-G002-17
Catalog No. 50-233-5433 Supplier Phenomenex Inc Supplier No. 7HKG00217
Zebron™ ZB-5 7HK-G002-17