Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc GC Cap. Column 30 m x 0.53 mm x 1.50 m

Catalog No. 502335442 Shop All Phenomenex Products
Encompass_Preferred

Zebron™ ZB-5 w/GUARDIAN™, 5 m 7HK-G002-28-GGA

Catalog No. 50-233-5442 Supplier Phenomenex Inc Supplier No. 7HKG00228GGA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zebron™ ZB-5 w/GUARDIAN™, 5 m 7HK-G002-28-GGA

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.