Learn More
Phenomenex Inc GC Cap. Column 50 m x 0.20 mm x 0.33 m
Catalog No. 502335476 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-5MS 7JE-G010-14
Catalog No. 50-233-5476 Supplier Phenomenex Inc Supplier No. 7JEG01014
Zebron™ ZB-5MS 7JE-G010-14