Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.32 mm x 0.25 m
Catalog No. 502335591 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-SemiVolatiles 7HM-G027-11
Catalog No. 50-233-5591 Supplier Phenomenex Inc Supplier No. 7HMG02711
Add to Cart
Add to Cart
Zebron™ ZB-SemiVolatiles 7HM-G027-11