Learn More
Phenomenex Inc GC Cap. Column 15 m x 0.53 mm x 0.15 m
Catalog No. 502335641 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-1 7EK-G001-05
Catalog No. 50-233-5641 Supplier Phenomenex Inc Supplier No. 7EKG00105
Zebron™ ZB-1 7EK-G001-05