Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.25 mm x 0.10 m
Catalog No. 502335445 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-35HT 7HG-G025-02
Catalog No. 50-233-5445 Supplier Phenomenex Inc Supplier No. 7HGG02502
Zebron™ ZB-35HT 7HG-G025-02