Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.53 mm x 1.0 m
Catalog No. 502335672 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-5 7HK-G002-22
Catalog No. 50-233-5672 Supplier Phenomenex Inc Supplier No. 7HKG00222
Zebron™ ZB-5 7HK-G002-22