Learn More
Phenomenex Inc Zebron ZB-5HT
Catalog No. 502335680 Shop All Phenomenex Products
Encompass_Preferred
Zebron ZB-5HT GC Cap. Column 15 m x 0.25 mm x 0.25 µm
Catalog No. 50-233-5680 Supplier Phenomenex Inc Supplier No. 7EGG01511
Zebron ZB-5HT GC Cap. Column 15 m x 0.25 mm x 0.25 µm