Learn More
Phenomenex Inc GC Cap. Column 30 m x 0.25 mm x 0.25 m
Catalog No. 502335435 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-1plus 7HG-G031-11
Catalog No. 50-233-5435 Supplier Phenomenex Inc Supplier No. 7HGG03111
Zebron™ ZB-1plus 7HG-G031-11