Learn More
Phenomenex Inc GC Cap. Column 60 m x 0.32 mm x 3.00 m
Catalog No. 502335522 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-1 7KM-G001-36
Catalog No. 50-233-5522 Supplier Phenomenex Inc Supplier No. 7KMG00136
Zebron™ ZB-1 7KM-G001-36