Learn More
Phenomenex Inc GC Cap. Column 100m x 0.25mm x 0.5m
Catalog No. 502335527 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-DHA-PONA 7MG-G042-17
Catalog No. 50-233-5527 Supplier Phenomenex Inc Supplier No. 7MGG04217
Zebron™ ZB-DHA-PONA 7MG-G042-17