Learn More
Phenomenex Inc GC Cap. Column 50 m x 0.25 mm x 0.25 m
Catalog No. 502335346 Shop All Phenomenex Products
Encompass_Preferred
Zebron™ ZB-WAX 7JG-G007-11
Catalog No. 50-233-5346 Supplier Phenomenex Inc Supplier No. 7JGG00711
Zebron™ ZB-WAX 7JG-G007-11