Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc Zebron PLUS Liner for Shimadzu 17A

Catalog No. 502339176 Shop All Phenomenex Products
Encompass_Preferred

Zebron PLUS Liner for Shimadzu 17A 3.4mm ID Straight Z-Liner™

Catalog No. 50-233-9176 Supplier Phenomenex Inc Supplier No. AG23B0301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zebron PLUS Liner for Shimadzu 17A 3.4mm ID Straight Z-Liner™

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.