Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 3.4mm ID Single Taper Z-Liner™

Supplier:  Phenomenex Inc AG13B1301

Encompass_Preferred

Zebron™ Liner for Shimadzu 17A AG1-3B13-01

Catalog No. 50-233-9144


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.