Learn More
Phenomenex Inc Phenomenex Guard Column, Zebron Z-Guard, Metal, 5 m, 0.53 mm
Catalog No. NC3628585 Shop All Phenomenex Products
Encompass
The Zebron Z-Guard metal guard column is a GC capillary guard designed to protect analytical columns by trapping nonvolatile residues and particulates, extending column lifetime and maintaining chromatographic performance.
  • Extends column lifetime by trapping nonvolatile residues and particulates.
  • Protects analytical capillary columns and preserves chromatographic performance.
  • Metal construction for robust connections and compatibility with GC capillary systems.
  • 5 m length with 0.53 mm internal diameter for standard GC configurations.
  • Intended as a replaceable guard to prevent stationary phase damage and improve peak shape.
Catalog No. NC3628585 Supplier Phenomenex Inc Supplier No. 7AKG00000GM0
The Zebron Z-Guard metal guard column is a GC capillary guard designed to protect analytical columns by trapping nonvolatile residues and particulates, extending column lifetime and maintaining chromatographic performance.
  • Extends column lifetime by trapping nonvolatile residues and particulates.
  • Protects analytical capillary columns and preserves chromatographic performance.
  • Metal construction for robust connections and compatibility with GC capillary systems.
  • 5 m length with 0.53 mm internal diameter for standard GC configurations.
  • Intended as a replaceable guard to prevent stationary phase damage and improve peak shape.