Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phenomenex Inc 2 Columns, 5m Guard and Y-Splitter

Catalog No. 502399823 Shop All Phenomenex Products
Encompass_Preferred

Zebron™ ZB-CLPesticides Kit 0.32mm ID KG0-9286

Catalog No. 50-239-9823 Supplier Phenomenex Inc Supplier No. KG09286
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zebron™ ZB-CLPesticides Kit 0.32mm ID KG0-9286

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.