Learn More
Enzo Life Sciences Zebularine (10mg). CAS: 3690-10-6
SDP
Supplier:  Enzo Life Sciences BMLGR3440010
Encompass_Preferred
Zebularine is a DNA methylation inhibitor which acts by forming a covalent complex with DNMT. It displays antiangiogenic activity and markedly inhibits vessel formation in different tumor models. It reactivates silenced E-cadherin and potentiates the effects of other epigenetic therapeutics such as vorinistat. Induces differentiation of mesenchymal cells into cardiomyocytes. Inhibits cytidine deaminase. Alternative name: 1-(beta-D-Ribofuranosyl)-1,2-dihydropyrimidin-2-one, 2-Pyrimidone-1-beta-D-riboside. Purity: ≥98% (TLC). Solubility: Soluble in water (>25 mg/ml) or DMSO (>25 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0999