Learn More
Apexbio Technology LLC Zebularine 3690-10-6 10mM (in 1mL DMSO)
SDP
Supplier:  Apexbio Technology LLC A19155.1
Encompass_Preferred
Zebularine (CAS number 3690-10-6) is a chemically stable cytidine analog containing a 2-(1H)-pyrimidinone ring structure It acts primarily through inhibition of DNA methylation by forming covalent complexes with DNA methyltransferases Zebularine treatment leads to demethylation and reactivation of hypermethylation-mediated silenced genes exemplified by the p16 gene Additionally induced myogenic differentiation has been observed upon zebularine administration in 10T1/2 cells Zebularine displays minimal cytotoxicity in certain cell models such as T24 cells and retains stability in aqueous solutions across a broad pH range (up to pH 12) These properties make zebularine relevant for studies in epigenetic regulation and cellular differentiation
Catalog No. 50-000-62854