Learn More
Abcam Zebularine, DNA methyltransferase (DNMT) inhibitor, 10MG
Catalog No. 502706746
Encompass_Preferred
MW 228.2 Da, Purity >99%. DNA methyltransferase( DNMT) inhibitor and cytidine deaminase inhibitor (Ki = 2 μM). Inhibits tumour growth in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6746 Supplier Abcam Supplier No. AB14126410MG
MW 228.2 Da, Purity >99%. DNA methyltransferase( DNMT) inhibitor and cytidine deaminase inhibitor (Ki = 2 μM). Inhibits tumour growth in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement