Learn More
Medchemexpress LLC Zectivimod | 1623066-63-6 | 99.35% | 528.47 | 50 MG
SDP
Catalog No. 5000351290
Encompass_Preferred
Zectivimod is a sphingosine-1-phosphate receptor agonist. It can be utilized in the research of autoimmune diseases, chronic inflammatory diseases, and immunoregulation disorders.
  • Sphingosine-1-phosphate receptor agonist
  • Research applications in autoimmune diseases
  • Research applications in chronic inflammatory diseases
  • Research applications in immunoregulation disorders
Catalog No. 50-003-51290 Supplier Medchemexpress LLC Supplier No. HY13955550MG
Zectivimod is a sphingosine-1-phosphate receptor agonist. It can be utilized in the research of autoimmune diseases, chronic inflammatory diseases, and immunoregulation disorders.
  • Sphingosine-1-phosphate receptor agonist
  • Research applications in autoimmune diseases
  • Research applications in chronic inflammatory diseases
  • Research applications in immunoregulation disorders