Learn More
Medchemexpress LLC Zectivimod | 1623066-63-6 | 99.4% | 528.5 g/mol | C28H31Cl2N3O3 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1395555MG
Encompass_Preferred
Zectivimod is a small-molecule sphingosine-1-phosphate receptor (S1P1) agonist used in research on immune modulation. It has been investigated as an orally available S1P1 modulator and is employed in preclinical and clinical research focused on autoimmune and chronic inflammatory diseases. The compound is supplied for laboratory research use only.
  • Selective S1P1 receptor agonist for immune modulation studies.
  • Investigated as an orally available S1P1 modulator.
  • Useful for research into autoimmune and chronic inflammatory diseases.
  • High chemical purity (>99%).
  • Supplied in small laboratory quantities for experimental use.
  • Not for human or veterinary use; for research only.
Catalog No. 50-002-21319