Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zectivimod | 1623066-63-6 | 99.4% | C28H31Cl2N3O3 | 10MG
SDP

Catalog No. 5000260339
Encompass_Preferred

Zectivimod is a sphingosine-1-phosphate (S1P) receptor agonist supplied for preclinical research into immune modulation, autoimmune disorders, and chronic inflammatory disease models. It is provided as a solid research chemical with solubility and formulation guidance for in vitro and in vivo studies.

  • Acts as a sphingosine-1-phosphate receptor agonist for immunomodulation studies.
  • High purity (≈99.4%) suitable for preclinical research.
  • Available in multiple small-scale pack sizes for dosing and formulation work.
  • Soluble in DMSO with explicit in vivo formulation protocols available.
  • Storage and stability guidance provided for solid and solvent conditions.

Catalog No. 50-002-60339 Supplier Medchemexpress LLC Supplier No. HY13955510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zectivimod is a sphingosine-1-phosphate (S1P) receptor agonist supplied for preclinical research into immune modulation, autoimmune disorders, and chronic inflammatory disease models. It is provided as a solid research chemical with solubility and formulation guidance for in vitro and in vivo studies.

  • Acts as a sphingosine-1-phosphate receptor agonist for immunomodulation studies.
  • High purity (≈99.4%) suitable for preclinical research.
  • Available in multiple small-scale pack sizes for dosing and formulation work.
  • Soluble in DMSO with explicit in vivo formulation protocols available.
  • Storage and stability guidance provided for solid and solvent conditions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.