Learn More
Medchemexpress LLC Zedoresertib (Debio 0123) | 2243882-74-6 | 99.5% | 511.45 | C26H28Cl2N6O | 5 MG
SDP
Catalog No. 5000222621
Encompass_Preferred
Zedoresertib (Debio 0123) is a potent, selective WEE1 kinase inhibitor used in preclinical research on cell-cycle regulation and DNA damage response. Supplied as a solid or as a DMSO solution, the compound is characterized by high purity and defined physicochemical properties to support reproducible laboratory experiments.
  • Potent WEE1 kinase inhibitor (IC50 0.8 nM).
  • High recommended purity of 99.46%.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Molecular weight 511.45 and formula C26H28Cl2N6O.
  • Intended for cell-cycle and DNA damage response studies.
  • Includes product datasheet and SDS for handling and safety information.
Catalog No. 50-002-22621 Supplier Medchemexpress LLC Supplier No. HY1470545MG
Zedoresertib (Debio 0123) is a potent, selective WEE1 kinase inhibitor used in preclinical research on cell-cycle regulation and DNA damage response. Supplied as a solid or as a DMSO solution, the compound is characterized by high purity and defined physicochemical properties to support reproducible laboratory experiments.
  • Potent WEE1 kinase inhibitor (IC50 0.8 nM).
  • High recommended purity of 99.46%.
  • Available as a solid and as a 10 mM solution in DMSO.
  • Molecular weight 511.45 and formula C26H28Cl2N6O.
  • Intended for cell-cycle and DNA damage response studies.
  • Includes product datasheet and SDS for handling and safety information.