Learn More
Medchemexpress LLC Zein | 9010-66-6 | >80.0% | 22-24 kDa | 25g
SDP
Catalog No. 5000269018
Encompass_Preferred
Zein is a plant protein based polymer can be used to prepare nanofibrous mats through electrospun Zein has good cell compatibility and easy fabrication ability and can be used in drug delivery systems[1]
Catalog No. 50-002-69018 Supplier Medchemexpress LLC Supplier No. HYW76380625G
Zein is a plant protein based polymer can be used to prepare nanofibrous mats through electrospun Zein has good cell compatibility and easy fabrication ability and can be used in drug delivery systems[1]