Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Zein, NF, Spectrum™ Chemical
SDP

Catalog No. 18607812 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-607-812 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-607-812 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. Z113125KGBL
Get Up to $300 in Additional Chemicals
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Z1131, 9010-66-6

Spectrum™ Chemical Zein, NF is used as a coating for pills and other encapsulated drugs. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9010-66-6
MDL Number MFCD00062414

Specifications

CAS Max % 1
Ignition Residue 0.02
Loss on Drying 0.08
Packaging Fiber Drum
Quantity 25 kg
Grade NF
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Zein
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.