Learn More
Hitech Instruments Inc Zeiss, Immersion Oil, 518F
SDP
Catalog No. NC2187819
Encompass
Zeiss Immersion Oil 518F is a high-quality immersion oil designed for microscopy. It has a refractive index of 1.518 at 23°C, making it suitable for objectives with high numerical apertures. The oil is free of fluorescence, has high transparency and is made of purified paraffin oil that will not damage microscope objectives. It can be used for both transmitted light and fluorescence microscopy.
This item is not returnable. View return policy
Catalog No. NC2187819 Supplier Hitech Instruments Inc Supplier No. 4338029010000
Zeiss Immersion Oil 518F is a high-quality immersion oil designed for microscopy. It has a refractive index of 1.518 at 23°C, making it suitable for objectives with high numerical apertures. The oil is free of fluorescence, has high transparency and is made of purified paraffin oil that will not damage microscope objectives. It can be used for both transmitted light and fluorescence microscopy.