Learn More
Selleck Chemical LLC Zelasudil E1978-1G
SDP
Catalog No. 5000039156
Encompass_Preferred
Zelasudil (RXC007) is a selective inhibitor of Rho-associated coiled-coil-containing kinase 2 (ROCK2) It has been found to have the antifibrotic effect
Catalog No. 50-000-39156 Supplier Selleck Chemical LLC Supplier No. E19781G
Zelasudil (RXC007) is a selective inhibitor of Rho-associated coiled-coil-containing kinase 2 (ROCK2) It has been found to have the antifibrotic effect