Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Zelavespib formic (PU-H71) | 00-00-0 | 97.4% | 558.39 g/mol | C19H23IN6O4S | 5 MG
SDP

Catalog No. 5000263344
Encompass_Preferred

Zelavespib formic is a research-grade small-molecule inhibitor of heat shock protein 90 (Hsp90), supplied as a formic acid salt for laboratory studies. It is intended for in vitro and preclinical research use only.

  • Potent Hsp90 inhibitor (IC50 51 nM in MDA-MB-468 cells).
  • Formic acid salt suitable for biological assays.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (97.39%).
  • Molecular weight 558.39 g/mol; formula C19H23IN6O4S.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-63344 Supplier Medchemexpress LLC Supplier No. HY11038D5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Zelavespib formic is a research-grade small-molecule inhibitor of heat shock protein 90 (Hsp90), supplied as a formic acid salt for laboratory studies. It is intended for in vitro and preclinical research use only.

  • Potent Hsp90 inhibitor (IC50 51 nM in MDA-MB-468 cells).
  • Formic acid salt suitable for biological assays.
  • Available as solid and as a 10 mM solution in DMSO.
  • High purity (97.39%).
  • Molecular weight 558.39 g/mol; formula C19H23IN6O4S.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.