Learn More
Medchemexpress LLC Zelavespib hydrochloride | 2095432-24-7 | 99.25% | 548.83 | 25 MG
SDP
Catalog No. 5000345639
Encompass_Preferred
Zelavespib hydrochloride is a potent Hsp90 inhibitor primarily used for research. It demonstrates significant activity against various tumor cells, inducing apoptosis and inhibiting growth.
  • Potent Hsp90 inhibitor with an IC50 of 51 nM in MDA-MB-468 cells.
  • Inhibits growth of multiple tumor cell lines.
  • Induces significant apoptosis in triple-negative breast cancers (TNBCs).
  • Downregulates oncoproteins involved in the invasive potential of TNBCs.
  • Cytotoxic to CLL cells with minimal effects on PBMC or resting B cells.
  • Reduces CLL viability by inducing mitochondrial apoptosis.
Catalog No. 50-003-45639 Supplier Medchemexpress LLC Supplier No. 25MGHY11038B
Add to Cart
Add to Cart
Zelavespib hydrochloride is a potent Hsp90 inhibitor primarily used for research. It demonstrates significant activity against various tumor cells, inducing apoptosis and inhibiting growth.
  • Potent Hsp90 inhibitor with an IC50 of 51 nM in MDA-MB-468 cells.
  • Inhibits growth of multiple tumor cell lines.
  • Induces significant apoptosis in triple-negative breast cancers (TNBCs).
  • Downregulates oncoproteins involved in the invasive potential of TNBCs.
  • Cytotoxic to CLL cells with minimal effects on PBMC or resting B cells.
  • Reduces CLL viability by inducing mitochondrial apoptosis.