Learn More
Selleck Chemical LLC Zelavespib10mg
SDP
Supplier:  Selleck Chemical LLC S803910MG
Encompass_Preferred
Zelavespib (PU-H71, NSC 750424) is a potent and selective inhibitor of HSP90 with IC50 of 51 nM. Phase 1. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3337