Learn More
AdipoGen Zelkovamycin (1 mg)
SDP
Supplier:  AdipoGen AGCN20128M001
Encompass_Preferred
Chemical. CAS: 221197-33-7. Formula: C36H45N9O9S. MW: 779.9. Isolated from Streptomyces sp. Cyclic peptide antibiotic Antibacterial.
Catalog No. 50-205-3501